EMAIL THIS PAGE TO A FRIEND
The Journal of organic chemistry
Reaction of O-methyl-N,N'-diisopropylisourea with amino acids and amines.
PMID 830852
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
66638
N,N′-Diisopropyl-O-methylisourea, purum, ≥97.0% (GC)
C8H18N2O