EMAIL THIS PAGE TO A FRIEND
Nature
Recognition sequence of specific endonuclease BamH.I from Bacillus amyloliquefaciens H.
PMID 834250
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
BAMHI-RO BamH I, from Bacillus amyloliquefaciens (H)