EMAIL THIS PAGE TO A FRIEND
The American journal of physiology
Purification and characterization of chlorotoxin, a chloride channel ligand from the venom of the scorpion.
PMID 8383429
Abstract
We have previously demonstrated that the venom of the scorpion Leiurus quinquestriatus blocks small-conductance Cl- channels, derived from epithelial cells, when applied to the cytoplasmic surface. We have now purified to near homogeneity, and characterized, the component responsible for this blocking activity. It is a small basic peptide of 4,070 Da. The primary amino acid structure shows considerable homology to a class of previously described putative short insectotoxins. A brief characterization of the kinetics of Cl- channel block as well as a demonstration of toxicity to arthropods is also presented.
Related Materials