EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Receptor-mediated transport of DNA into eukaryotic cells.
PMID 8386298
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N1127
2-Nitrophenyl β-D-galactopyranoside, ≥98% (enzymatic)
C12H15NO8