EMAIL THIS PAGE TO A FRIEND
Journal of bacteriology
Deoxyribonucleic acid polymerase from the extreme thermophile Thermus aquaticus.
PMID 8432
Abstract
A stable deoxyribonucleic acid (DNA) polymerase (EC 2.7.7.7) with a temperature optimum of 80 degrees C has been purified from the extreme thermophile Thermus aquaticus. The enzyme is free from phosphomonoesterase, phosphodiesterase and single-stranded exonuclease activities. Maximal activity of the enzyme requires all four deoxyribonucleotides and activated calf thymus DNA. An absolute requirement for divalent cation cofactor was satisfied by Mg2+ or to a lesser extent by Mn2+. Monovalent cations at concentrations as high as 0.1 M did not show a significant inhibitory effect. The pH optimum was 8.0 in tris(hydroxymethyl)aminomethane-hydrochloride buffer. The molecular weight of the enzyme was estimated by sucrose gradient centrifugation and gel filtrations on Sephadex G-100 to be approximately 63,000 to 68,000. The elevated temperature requirement, small size, and lack of nuclease activity distinguish this polymerase from the DNA polymerase of Escherichia coli.