EMAIL THIS PAGE TO A FRIEND
Journal of cardiothoracic and vascular anesthesia
Use of amrinone in cardiac surgery patients.
PMID 8471743
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4056
Amrinone
C10H9N3O