EMAIL THIS PAGE TO A FRIEND
Revue francaise de gynecologie et d'obstetrique
[Rupture of an unscarred uterus at full term after an intracervical application of dinoprostone (Prepidil) gel].
PMID 8493447
Abstract
The authors report a case of uterine rupture following intra-cervical application of dinoprostone (two doses at an interval of 6 hours). No oxytocic had been administered and recording of the contractions had revealed no hyperkinesia or hypertonia one hour before rupture occurred.