EMAIL THIS PAGE TO A FRIEND
Journal of chromatography
Ion-pair extraction of histamine from biological fluids and tissues for its determination by high-performance liquid chromatography with fluorescence detection.
PMID 8496284
Abstract
A procedure for the selective ion-pair extraction of histamine from various biological fluids and tissues by using the ion-pairing agent bis(2-ethylhexyl)hydrogen-phosphoric acid has been developed. This procedure enabled routine determination of histamine in plasma, urine and tissue samples of humans and rats by reversed-phase high-performance liquid chromatography with fluorescence detection using o-phthaldialdehyde.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
53300
Histamine dihydrochloride, ≥99.0% (AT)
C5H9N3 · 2HCl