EMAIL THIS PAGE TO A FRIEND
International journal of fertility and menopausal studies
Norethindrone--a review of therapeutic applications.
PMID 8520623
Abstract
Norethindrone is utilized for numerous noncontraceptive therapies, especially during the menopause. Through an analysis of the literature we have described the pharmacological profile of norethindrone and the potential therapeutic implications of low-dose therapies, with an emphasis on endometrial pathology, climacteric symptoms, lipid metabolism, and bone density.