EMAIL THIS PAGE TO A FRIEND
Psychopharmacology
D4 dopamine receptor binding affinity does not distinguish between typical and atypical antipsychotic drugs.
PMID 8524985
Abstract
The affinities of 13 atypical and 12 typical antipsychotic drugs for the cloned rat D4 dopamine receptor and the D4/D2 ratios were examined. Of the atypical antipsychotic drugs tested, only clozapine, risperidone, olanzapine, zotepine and tiospirone had affinities less than 20 nM. In fact, many atypical antipsychotic drugs had relatively low affinities for the cloned rat D4 receptor, with Ki values greater than 100 nM (Seroquel, fluperlapine, tenilapine, FG5803 and melperone). Additionally, several typical antipsychotic drugs had high affinities for the cloned rat D4 receptor, with Kis less than 20 nM (loxapine, chlorpromazine, fluphenazine, mesoridazine, thioridazine and trifluoroperazine). The ratios of D2/D4 affinities did not differentiate between these two types of antipsychotic drugs. Thus, D4 dopamine receptor affinity, used as a single measure, does not distinguish between the group of typical and atypical antipsychotic drugs analyzed.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
L106
Loxapine succinate salt, solid
C18H18ClN3O · C4H6O4