EMAIL THIS PAGE TO A FRIEND
Progress in clinical and biological research
The species specificity of dieldrin- or phenobarbital- induced hepatocarcinogenesis: case studies with implications for human health risk assessment.
PMID 8532726