EMAIL THIS PAGE TO A FRIEND
The British journal of dermatology
Aplasia cutis congenita after exposure to methimazole: a causal relationship?
PMID 8547058
Abstract
We report a child with scalp aplasia cutis congenita, whose mother was treated with methimazole during pregnancy. The relationship between antithyroid drug administration during pregnancy and the occurrence of scalp defects is discussed, and the pertinent literature is reviewed.