EMAIL THIS PAGE TO A FRIEND
British journal of pharmacology
Pharmacological characterization of two novel and potent 5-HT4 receptor agonists, RS 67333 and RS 67506, in vitro and in vivo.
PMID 8564196
Abstract
1. The pharmacology of two novel 5-HT4 receptor agonists, RS 67333 (1-(4-amino-5-chloro-2-methoxy-phenyl)-3-[1(n-butyl)-4-piperidinyl]-1- propanone HCl) and RS 67506 (1-(4-amino-5-chloro-2-methoxy-phenyl)-3-[1-(2-methyl sulphonylamino)ethyl-4-piperidinyl]-1-propanone HCl) have been assessed in vitro and in vivo. 2. RS 67333 and RS 67506 exhibited affinities (pKi = 8.7 and 8.8, respectively) for the 5-HT4 binding sites, labelled with [3H]-GR 113808, in guinea-pig striatum. The Hill coefficients from these displacement curves were not significantly different from unity. The compounds exhibited lower affinities (< 6.0) at several other receptors including 5-HT1A, 5-HT1D, 5-HT2A, 5-HT2C, dopamine D1, D2 and muscarinic M1-M3 receptors. However, RS 67333 and RS 67506 did exhibit affinities for the sigma 1 (pKi = 8.9 and 7.9, respectively) and sigma 2 (pKi = 8.0 and 7.3, respectively) binding sites. 3. At the 5-HT4 receptor mediating relaxation of the carbachol-precontracted oesophagus, RS 67333 and RS 67506 acted as potent (pEC50 8.4 and 8.6, respectively), partial agonists (intrinsic activities, with respect to 5-HT were 0.5 and 0.6, respectively) with respect to 5-HT. Relaxant responses to RS 67333 or RS 67506 were surmountably antagonized by GR 11308 (10 nM), with apparent affinities (pKB) of 9.1 and 9.0, respectively. RS 67333 and RS 67506 induced dose-dependent increases in heart rate of the anaesthetized micropig (ED50 4.9 and 5.4 micrograms kg-1, i.v.), with maximal increases of 35 and 47 beats min-1, respectively. 4. RS 67333 and RS 67506, therefore, acted as potent, partial 5-HT4 receptor agonists in vitro and in vivo. These compounds, by virtue of their high potency and selectivity, may have some utility in elucidating the physiological role of 5-HT4 receptors.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
SML1882
RS 67333 hydrochloride, ≥98% (HPLC)
C19H29ClN2O2 ·HCl