EMAIL THIS PAGE TO A FRIEND
Journal of chemotherapy (Florence, Italy)
The place of tobramycin in lower respiratory tract infections (LRTI).
PMID 8568545
Abstract
The Author provides a review of clinical experience with tobramycin as therapy for lower respiratory tract infections, in comparison to other aminoglycosides, including the pharmacokinetics and toxicity, dwelling on oto- and nephrotoxicity. The article includes a discussion of various dosing regimens of the aminoglycosides, focussing on efficacy and toxicity arising from once-daily administration. The Author then provides a more detailed description of tobramycin's pharmacokinetics, indications for its use, and the possibilities of once-daily dosing, concluding that toxicity is favorably influenced by a single daily administration as well as efficacy, and that patient compliance and reduced hospital costs are other advantages of this regimen.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001210
Tobramycin for identification, European Pharmacopoeia (EP) Reference Standard
C18H37N5O9