EMAIL THIS PAGE TO A FRIEND
The Journal of pharmacology and experimental therapeutics
Effects of benztropine on behavioral and toxic effects of cocaine: comparison with atropine and the selective dopamine uptake inhibitor 1-[2-(diphenylmethoxy)ethyl]-4-(3-phenyl-propyl)-piperazine.
PMID 8613919
Abstract
Behavioral effects of cocaine that are relevant to its abuse have been associated with pharmacological actions at the dopamine uptake carrier. Benztropine (Cogentin) is an antiparkinson agent that has limited abuse despite its ability to block dopamine uptake, and has been suggested as a candidate for the treatment of cocaine dependence. Preclinical studies were conducted to assess the behavioral and toxic effects of benztropine alone and in conjunction with cocaine. Because of the mixed pharmacology of benztropine which includes antimuscarinic as well as dopaminergic actions, results obtained from parallel experiments with atropine and the selective dopamine uptake inhibitor, GBR 12935 (1-[2-(diphenylmethoxy)ethyl]-4-(3-phenyl-propyl)-piperazine), were performed. All of the drugs stimulated locomotor activity of mice, but atropine and benztropine had much lower efficacy. Nonstimulatory doses of GBR 12935 enhanced the locomotor stimulant effects of cocaine, whereas benztropine and atropine did not share this effect. GBR 12935, benztropine and cocaine increased fixed-interval responding, whereas atropine decreased fixed-interval response rates in rats. Only GBR 12935 and cocaine increased responding during timeout periods. GBR 12935, but not benztropine or atropine, fully reproduced the discriminative stimulus effects of cocaine (10 mg/kg). GBR 12935 and atropine augmented the discriminative stimulus effects of lower cocaine doses in rats. Only GBR 12935 and cocaine had convulsant effects and only GBR 12935 significantly enhanced the convulsant effects of cocaine in mice. These results document a behavioral and toxicity profile for benztropine distinct from that of classical dopamine uptake blockers. The data underscore further the potential of benztropine as a candidate for clinical evaluation in the treatment of cocaine dependence.