EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Posttranslational regulation of a Leishmania HEXXH metalloprotease (gp63). The effects of site-specific mutagenesis of catalytic, zinc binding, N-glycosylation, and glycosyl phosphatidylinositol addition sites on N-terminal end cleavage, intracellular stability, and extracellular exit.
PMID 8626468
Abstract
Leishmanolysin (EC 3.4.24.36) (gp63) is a HEXXH metalloprotease, encoded by multicopied genes in Leishmania and implicated in the infectivity of these parasitic protozoa. We examined posttranslational regulation of gp63 expression by site-specific mutagenesis of the predicted catalytic/zinc-binding sites in the H264EXXH motif, the potential sites of N-glycosylation and glycosyl phosphatidylinositol addition. Mutant and wild-type genes were cloned into a Leishmania-specific vector for transfecting a deficient variant, which produced gp63 approximately 20-fold less than wild-type cells. The selective conditions chosen fully restored this deficiency in transfectants with the wild-type gene. Under these conditions, all transfectants were found comparable in both the plasmid copy number per cell and elevation of gp63 transcripts. Mutant and wild-type products in the transfectants were then compared quantitatively and qualitatively by specific immunologic and protease assays. The results indicate the following. 1) Glu-265 in the HEXXH motif is indispensable for the catalytic activity of gp63. The propeptide of the inactive mutant products was cleaved, suggestive of a non-intramolecular event. 2) Substitution of either His residue in HEXXH leads to apparent intracellular degradation of the mutant products, pointing to a role for zinc binding in in vivo stability of gp63. 3) The three potential sites of N-glycosylation at Asn-300, Asn-407, and Asn-534 are all utilized and contribute to intracellular stability of gp63. 4) Substitution of Asn-577 causes release of all mutant products, indicative of its specificity as a glycosyl phosphatidylinositol addition site for membrane anchoring of gp63. It is suggested that expression of gp63 as a functional protease is regulated by these posttranslational modification pathways.