EMAIL THIS PAGE TO A FRIEND
The American journal of the medical sciences
Milrinone: basic and clinical pharmacology and acute and chronic management.
PMID 8659556
Abstract
Milrinone (Inocor-Sanofi-Winthrop) represents a second generation phosphodiesterase inhibitor currently approved for intravenous administration in the treatment of decompensated congestive heart failure. By inhibiting Type III phosphodiesterase, milrinone increases intracellular cyclic adenosine monophosphate. This results in a positive inotropic effect on the heart and vasodilatation in the periphery. The hemodynamic consequences of this action produce left ventricular afterload reduction, with an increase in cardiac output and a reduction in total peripheral resistance. Unlike the sympathomimetic amines, milrinone produces no tolerance and possesses the distinct advantage of directly decreasing pulmonary vascular resistance. Short-term intermittent infusion by peripheral administration, continuous infusion, long-term therapy, and intermittent outpatient therapy was demonstrated to be safe, efficacious, and cost effective. It is hypothesized that intravenous milrinone, by producing biventricular afterload reduction, offers an efficacious, cost-effective tool for the treatment of decompensated heart failure.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
M4659
Milrinone, ≥97% (TLC), powder
C12H9N3O