EMAIL THIS PAGE TO A FRIEND
Pharmacological research communications
Adriamycin associated cardiotoxicity: research on prevention with coenzyme Q.
PMID 866369
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
27597
Coenzyme Q9, ≥96.0% (HPLC)
C54H82O4