EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Molecular characterization of AKAP149, a novel A kinase anchor protein with a KH domain.
PMID 8769136
Abstract
The cytosolic cAMP activates in eukaryotic cells several isoforms of cAMP-dependent protein kinase (PKAs) involved in signal transduction. The effects of individual PKA isoforms are determined by their cellular localisation, specified through binding to distinct A Kinase Anchor Proteins (AKAPs). A new member of the AKAP family, a membrane-anchored 903 amino acid long protein, designated AKAP149, is characterized in the present work. It is a putative splicing variant of S-AKAP84 with the important new feature of a RNA-binding motif (KH domain). This domain together with the known characteristics of AKAPs suggests the involvement of AKAP149 in the phosphorylation-dependent regulation of RNA-processing.