EMAIL THIS PAGE TO A FRIEND
Methods in enzymology
Nitric oxide synthase inhibitors: amino acids.
PMID 8782604
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
72750
Nω-Nitro-L-arginine, ≥99.0% (NT)
C6H13N5O4