EMAIL THIS PAGE TO A FRIEND
Genomics
Chromosomal mapping of the gene (INPP5A) encoding the 43-kDa membrane-associated inositol polyphosphate 5-phosphatase to 10q26.3 by fluorescence in situ hybridization.
PMID 8808294