EMAIL THIS PAGE TO A FRIEND
Blood
Filgrastim (r-metHuG-CSF): the first 10 years.
PMID 8822908
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001173 Filgrastim, European Pharmacopoeia (EP) Reference Standard