EMAIL THIS PAGE TO A FRIEND
Biochimie
Induction of apoptotic cell death by DNA topoisomerase II inhibitors.
PMID 8824770
Abstract
We have analyzed the interference of antitumoral drugs acting through the inhibition of DNA topoisomerase II on the human HeLa cell metabolism. Different compounds characterized by a diverse mechanism of action have been used, namely m-amsacrine, an intercalative drug, etoposide, which does not intercalate DNA, and suramin, which exerts its effect through an unknown mechanism. In HeLa cells treated with increasing doses of these drugs, we have examined cell viability and DNA synthesis capacity, and we have evaluated topoisomerase II activity. Cellular morphology and DNA integrity have been studied in order to characterize the mechanism of cell death. The results we have obtained clearly indicate that topoisomerase II poisons induce cell death by apoptosis. These observations suggest a role of the inhibition of topoisomerase II activity in the apoptotic program.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9809
Amsacrine hydrochloride, ≥98% (TLC), powder
C21H19N3O3S · HCl