EMAIL THIS PAGE TO A FRIEND
The Annals of pharmacotherapy
Comparison of cefprozil, cefpodoxime proxetil, loracarbef, cefixime, and ceftibuten.
PMID 8833562
Abstract
To discuss the pharmacokinetics, spectrum of activity, clinical trials, and adverse effects of cefprozil, cefpodoxime proxetil, loracarbef, cefixime, and ceftibuten, an investigational cephalosporin. Literature was identified by a MEDLINE search from 1986 to January 1995. Randomized, controlled studies were selected for evaluation; however, uncontrolled studies were included when data were limited for indications approved by the Food and Drug Administration. Data were evaluated with respect to in vitro activity, study design, clinical and microbiologic outcomes, and adverse drug reactions. Cefprozil, cefpodoxime proxetil, loracarbef, cefixime, and cefributen are active in vitro against organisms frequently involved in community-acquired infections such as Streptococcus pneumoniae, Escherichia coli, beta-lactamase-positive or -negative Haemophilus influenzae, and Moraxella catarrhalis. Except for cefixime and ceflibuten, they all are active against methicillin-susceptible Staphylococcus aureus. Even though there were problems in study design (discussed within the text), clinical data demonstrate that these new oral beta-lactam compounds are as efficacious as conventional therapies for a variety of community-acquired infections. Cefprozil, cefpodoxime, cefixime, loracarbef, and ceftibuten demonstrate in vitro activity against the major organisms that cause community-acquired infections. Clinical trials confirm that these agents are as effective as traditional therapies for the management of acute otitis media, pharyngitis/tonsillitis, sinusitis, bronchitis, pneumonia, urinary tract infections, and skin and skin-structure infections. In addition, cefixime and cefpodoxime are effective therapies for uncomplicated gonococcal infections. Selection of a specific agent will be influenced by susceptibility data and safety, as well as issues of compliance and cost.