EMAIL THIS PAGE TO A FRIEND
Histochemistry and cell biology
Evaluation of five green fluorescence-emitting streptavidin-conjugated fluorochromes for use in immunofluorescence microscopy.
PMID 8877388
Abstract
Fluorescein isothiocyanate (FITC) is largely used in immunofluorescence methods. We propose to analyse the quality of some recent fluorochromes using image analysis. Fluorochromes tested include FITC and dichlorotriazinylaminofluorescein (DTAF), dipyrrometheneboron difluoride (BODIPY), Rhodol Green and cyanine 2. RAMOS cells were immunolabelled against the proliferating cell nuclear antigen (PCNA) revealed by the biotin-streptavidin technique. Slides were mounted in anhydrous glycerol or in buffered glycerol (pH 7.0 or pH 8.5). No antifading medium was added. Cell fluorescence emission intensity and bleaching characteristics were measured. Rhodol Green exhibited the highest fluorescence intensity and the best photobleaching resistance. Although BODIPY also resisted well during the photobleaching assay, its fluorescence intensity was weak. FITC, DTAF and cyanine 2 showed intermediate fluorescence intensity and a fast decay of fluorescence. Among the green emitting fluorochromes tested, Rhodol Green appeared to be the best.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
67806
Rhodol, ≥85% (HPCE)
C20H13NO4