EMAIL THIS PAGE TO A FRIEND
Electrophoresis
Biotinylated hyaluronan: a versatile and highly sensitive probe capable of detecting nanogram levels of hyaluronan binding proteins (hyaladherins) on electroblots by a novel affinity detection procedure.
PMID 8907541
Abstract
Hyaluronan influences cellular proliferation and migration in developing, regenerating and remodelling tissues and in tissues undergoing malignant tumour-cell invasion. The widespread occurrence of hyaluronan-binding proteins indicates that the recognition of hyaluronan is important to tissue organisation and the control of cellular behaviour. A number of extracellular matrix and cellular proteins, which have been termed the hyaladherins, have specific affinities for hyaluronan. These include cartilage link-protein, hyaluronectin, neurocan, versican and aggrecan, which all bind to HA within the extracellular matrix. Cellular receptors for hyaluronan such as CD44 and RHAMM (receptor for hyaluronate-mediated motility) have also been identified. In the present study biotinylated hyaluronan (bHA) was prepared by reacting adipic dihydrazide with a 170 kDa hyaluronan sample using the bifunctional reagent 1-ethyl-3-[3-(dimethylamino) propyl] carbodiimide. The resultant free amine moeity of the hydrazido-hyaluronan was then reacted with biotin succinimidyl ester (sulfo-NHS-biotin) to prepare the bHA. After 4-20% gradient sodium dodecyl sulfate-polyacrylamide gel electrophoresis (SDS-PAGE) and electroblotting to nitrocellulose membranes, bHA and avidin alkaline phosphatase conjugate could be used in conjunction with nitroblue tetrazolium/5-bromo-4-chloro-3-indolyl phosphate substrates to specifically visualise with high sensitivity (> or = 2 ng), bovine nasal cartilage link-protein, aggrecan hyaluronan binding region, and human fibroblast hyaluronan receptors such as CD-44. Conventional Western blotting using specific monoclonal antibodies to these proteins was also used to confirm the identities of these proteins.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
H9910 Hyaluronic Acid Binding Protein−Biotin bovine, lyophilized powder