EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Proteins containing reductively aminated disaccharides. Synthesis and chemical characterization.
PMID 900934
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5783 Albumin, lactosyl bovine, lyophilized powder