EMAIL THIS PAGE TO A FRIEND
Gastroenterology nursing : the official journal of the Society of Gastroenterology Nurses and Associates
Common vitamin B complex agents and folic acid: Part I. Cyanocobalamin (vitamin B12) (kaybovite [parenteral]; berubigen; betalin-12; kaybovite-1000; rubramin-pc).
PMID 9025404