EMAIL THIS PAGE TO A FRIEND
Indian journal of biochemistry & biophysics
Gramicidin channels--a solvable membrane "protein" folding problem.
PMID 9029812
Abstract
The linear gramicidins are peptide antibiotics that form cation-selective channels in lipid bilayers. Gramicidin channels have very well-defined functional characteristics, and the structure of membrane-spanning gramicidin A channels is known at atomic resolution. These features make the gramicidins well suited to study how the amino acid sequence encodes the structure and function of a membrane-spanning channel. We show how one can use electrophysiological measurements to obtain structural information about conducting channels and to quantify the conformational preferences of sequence-substituted gramicidin mutants.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G0550000 Gramicidin