EMAIL THIS PAGE TO A FRIEND
American journal of hematology
Danazol for paroxysmal nocturnal hemoglobinuria.
PMID 9034290
Abstract
Paroxysmal nocturnal hemoglobinuria (PNH) is a rare clonal stem-cell disorder in which blood cells lack complement inhibiting membrane proteins, and become susceptible to complement-mediated injury, leading to chronic intravascular hemolysis and pancytopenia. Glucocorticoids have been a mainstay of therapy. For patients refractory to glucocorticoids and requiring blood transfusions, an alternative therapy is needed. We studied danazol therapy in 5 patients refractory to other treatments. Four of the 5 benefited, showing rise in hematocrit and eventual cessation of transfusion requirements. Remissions lasted > or =2 years in 3 and 10 years in 1 patient. Danazol was well-tolerated without serious side effects. Danazol appears to be a good alternative treatment in PNH.