EMAIL THIS PAGE TO A FRIEND
Chemical & pharmaceutical bulletin
Chemical and biochemical studies on carbohydrate esters. V. Anti Ehrlich ascites tumor effect and chromatographic behaviors of fatty acyl monoesters of sucrose and trehalose.
PMID 908095