EMAIL THIS PAGE TO A FRIEND
Biochemical and biophysical research communications
Role of Na+/Ca2+ exchanger in preventing Na+ overload and hepatocyte injury: opposite effects of extracellular and intracellular Ca2+ chelation.
PMID 9125111
Abstract
We have previously shown that an increase of intracellular Na+ occurs in isolated rat hepatocytes undergoing ATP depletion and that Na+ accumulation is associated with an uncontrolled influx of Ca2+ through the activation in reverse mode of the Na+/Ca2+ exchanger. In the present study we have investigated the relationship between alterations of Na+ and Ca2+ homeostasis and hepatocyte killing using treatments which differentially chelate extracellular or intracellular Ca2+. Chelation of extracellular Ca2+ by ethylene glycol bis-(beta-aminoethyl ether) N,N,N',N'-tetraacetic acid (EGTA) potentiated Na+ overload and cell killing induced in isolated rat hepatocytes by hypoxia or menadione. Similar effects were also observed when Na+ accumulation was induced by the combined addition of Na+ ionophore monensin and the inhibition of plasma membrane Na+/K+ ATPase by ouabain. Conversely, the use of the intracellular Ca2+ chelator EGTA acetoxymethyl ester (EGTA/AM) reduced Na+ overload and hepatocyte death induced by hypoxia or cell treatment with menadione or monensin plus ouabain. The effects of EGTA/AM were reverted in the presence of bepridil, an inhibitor of Na+/Ca2+ exchanger. Altogether these results indicated that differential chelation of intracellular or extracellular Ca2+ influences in opposite ways hepatocyte killing due to ATP depletion by modulating intracellular Na+ levels through the reversed activity of the Na+/Ca2+ exchanger.