EMAIL THIS PAGE TO A FRIEND
Nature
gamma endorphin, alpha endorphin and Met-enkephalin are formed extracellularly from lipotropin C fragment.
PMID 917112
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
E6136
α-Endorphin human, ≥97% (HPLC)
C77H120N18O26S