EMAIL THIS PAGE TO A FRIEND
Antimicrobial agents and chemotherapy
Antifungal pharmacodynamic characteristics of fluconazole and amphotericin B tested against Candida albicans.
PMID 9174207
Abstract
Time-kill curves were determined for three isolates of Candida albicans tested against fluconazole and amphotericin B at multiples of the MIC. Fluconazole produced fungistatic activity, with concentration-related growth effects observed over a narrow range of concentrations. Amphotericin B exhibited fungicidal activity, with enhancement of activity over a broader range of concentrations.