EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
The anti-angiogenic agent fumagillin covalently binds and inhibits the methionine aminopeptidase, MetAP-2.
PMID 9177176
Abstract
The inhibition of new blood vessel formation (angiogenesis) is an effective means of limiting both the size and metastasis of solid tumors. The leading anti-angiogenic compound, TNP-470, has proven to be effective in in vitro and in animal model studies, and is currently being tested in phase III antitumor clinical trials. Despite many detailed pharmacological studies, little is known of the molecular mode of action of TNP-470. Using a derivative of the TNP-470 parent compound, the fungal metabolite, fumagillin, we have purified a mammalian protein that is selectively and covalently bound by this natural product. This fumagillin binding protein was found to be a metalloprotease, methionine aminopeptidase (MetAP-2), that is highly conserved between human and Saccharomyces cerevisiae. In the absence of MetAP-1, a distantly related methionine aminopeptidase, MetAP-2 function is essential for vegetative growth in yeast. We demonstrate that fumagillin selectively inhibits the S. cerevisiae MetAP-2 protein in vivo. The binding is highly specific as judged by the failure of fumagillin to inhibit MetAP-1 in vivo. Hence, these results identify MetAP-2 as an important target of study in the analysis of the potent biological activities of fumagillin.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F6771
Fumagillin from Aspergillus fumigatus, ≥90%, powder
C26H34O7