EMAIL THIS PAGE TO A FRIEND
The cancer journal from Scientific American
Bicalutamide ('Casodex') development: from theory to therapy.
PMID 9263623
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
Y0001448
Bicalutamide for system suitability, European Pharmacopoeia (EP) Reference Standard
C18H14F4N2O4S