EMAIL THIS PAGE TO A FRIEND
The Pediatric infectious disease journal
Generic acyclovir vs. famciclovir and valacyclovir.
PMID 9306476
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F7932
Famciclovir, ≥98% (HPLC)
C14H19N5O4