EMAIL THIS PAGE TO A FRIEND
Cellular and molecular life sciences : CMLS
Regulation of cyclooxygenase enzymes by nitric oxide.
PMID 9312403
Abstract
Nitric oxide (NO), derived from L-arginine (L-Arg) by the enzyme nitric oxide synthase (NOS) is involved in the regulation of several important physiological and pathophysiological functions. The mechanisms by which NO exerts some of its beneficial or detrimental effects include activation of guanylate cyclase, formation of peroxynitrite, apoptosis, and regulation of cyclooxygenase (COX). Cyclooxygenase (COX) is the enzyme that converts arachidonic acid to prostaglandins (PG), prostacyclin (PGI2) and thromboxane A2. The role of NO in the regulation of COX and its importance in physiology, pathology and therapy will be reviewed. Evidence will be presented to suggest that COX enzymes are targets for the physiopathological roles of NO and that once activated in the presence of NO, they represent important transduction mechanisms for its multifaceted actions.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
P6188
Prostaglandin I2 sodium salt, ≥96% (HPLC), synthetic, powder
C20H31NaO5