EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
A classification of nucleotide-diphospho-sugar glycosyltransferases based on amino acid sequence similarities.
PMID 9334165
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
G5507 Galactosyltransferase from bovine milk, lyophilized powder