EMAIL THIS PAGE TO A FRIEND
Pharmacological research
Separation of DL-dopa by means of micellar electrokinetic capillary chromatography after derivatization with Marfey's reagent.
PMID 9356208
Abstract
Micellar electrokinetic capillary chromatography was used to separate levodopa (L-3,4-dihydroxyphenyl-L-alanine), dextrodopa (D-3,4-dihydroxyphenyl-L-alanine) and DL-3-O-methyl-dopa, a metabolite of dopa, which were derivatised with the Marfey's reagent, in sodium borate, sodium dodecyl sulfate and acetonitile buffer system. Whereas DL-dopa derivatives could be separated, DL-3-O-methyl-dopa derivatives could not.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D9503
3,4-Dihydroxy-DL-phenylalanine
C9H11NO4