EMAIL THIS PAGE TO A FRIEND
Immunology today
CD6-ligand interactions: a paradigm for SRCR domain function?
PMID 9357143
Abstract
The scavenger receptor cysteine-rich (SRCR) superfamily, which includes proteins expressed by leukocytes, can be subdivided into groups A and B. Group B contains the lymphocyte cell-surface receptor CD6. This article reviews recent progress in understanding the interaction between CD6 and its ligand, activated leukocyte cell adhesion molecule (ALCAM). Analysis of the CD6-ALCAM interaction may help to understand how other SRCR domains bind to their ligands.