EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
N,N'-carbonyldiimidazole-induced peptide formation in aqueous solution.
PMID 938667
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
705284
1,1′-Carbonyldiimidazole solution, 0.4 M in methylene chloride
C7H6N4O