Description
Molecular Formula
Add to Cart
93784
Caffeine, PharmaGrade, EP, Manufactured under appropriate GMP controls for pharma or biopharmaceutical production
C8H10N4O2
27602
Caffeine, anhydrous, tested according to Ph.Eur.
C8H10N4O2
C0750
Caffeine, powder, ReagentPlus®
C8H10N4O2
C8960
Caffeine, BioXtra
C8H10N4O2
C1778
Caffeine, Sigma Reference Standard, vial of 250 mg
C8H10N4O2
BP766
Caffeine, British Pharmacopoeia (BP) Reference Standard
C8H10N4O2
C0100000
Caffeine, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
C7731
Caffeine, meets USP testing specifications, anhydrous
C8H10N4O2
1085003
Caffeine, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1009
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
56396
Caffeine, certified reference material, TraceCERT®
C8H10N4O2
W222402
Caffeine, anhydrous, 99%, FCC, FG
C8H10N4O2
05-0370
Caffeine, SAJ special grade, ≥98.5%
C8H10N4O2
Y0000787
Caffeine for system suitability, European Pharmacopoeia (EP) Reference Standard
C8H10N4O2
1086006
Caffeine melting point standard, United States Pharmacopeia (USP) Reference Standard
C8H10N4O2
PHR1095
Caffeine Melting Point Standard, Pharmaceutical Secondary Standard; Certified Reference Material
C8H10N4O2
41019
Melting point standard 235-237°C, analytical standard
C8H10N4O2
75035
Mettler-Toledo Calibration substance ME 18872, Caffeine, analytical standard, for the calibration of the thermosystem 900, traceable to primary standards (LGC)
C8H10N4O2