EMAIL THIS PAGE TO A FRIEND
Analytical Biochemistry
A rapid and sensitive method for the quantitation of microgram quantities of protein utilizing the principle of protein-dye binding.
PMID 942051
Abstract
We're sorry this abstract is currently not available.
Set your institution to read full text papers when available.