EMAIL THIS PAGE TO A FRIEND
Immunity
MyD88: an adapter that recruits IRAK to the IL-1 receptor complex.
PMID 9430229
Abstract
IL-1 is a proinflammatory cytokine that signals through a receptor complex of two different transmembrane chains to generate multiple cellular responses, including activation of the transcription factor NF-kappaB. Here we show that MyD88, a previously described protein of unknown function, is recruited to the IL-1 receptor complex following IL-1 stimulation. MyD88 binds to both IRAK (IL-1 receptor-associated kinase) and the heterocomplex (the signaling complex) of the two receptor chains and thereby mediates the association of IRAK with the receptor. Ectopic expression of MyD88 or its death domain-containing N-terminus activates NF-kappaB. The C-terminus of MyD88 interacts with the IL-1 receptor and blocks NF-kappaB activation induced by IL-1, but not by TNF. Thus, MyD88 plays the same role in IL-1 signaling as TRADD and Tube do in TNF and Toll pathways, respectively: it couples a serine/threonine protein kinase to the receptor complex.