EMAIL THIS PAGE TO A FRIEND
Advance for nurse practitioners
Daypro (oxaprozin) offers advantages in arthritis.
PMID 9459947
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O9637
Oxaprozin, solid
C18H15NO3
1482207
Oxaprozin, United States Pharmacopeia (USP) Reference Standard
C18H15NO3