EMAIL THIS PAGE TO A FRIEND
Anti-cancer drug design
Antiproliferative activity of colchicine analogues on MDR-positive and MDR-negative human cancer cell lines.
PMID 9474240
Abstract
In this study the in vitro antitumor activity of a series of 20 colchicine analogues was tested and compared with colchicine and thiocolchicine on three different human cancer cell lines, two of which express the multidrug-resistance (MDR) phenotype. At concentrations from 1 nM to 100 microM, all compounds tested inhibited cancer cell proliferation. The IC50 values indicate that the three fluorinated analogues were the most active compounds, with a similar decreasing order of potency (IDN 5005 > IDN 5079 > IDN 5080) on the two MDR-expressing cell lines, whereas thiocolchicine was the most effective compound on the MDR-negative MDA-MB 231 cells. A strong correlation (r = 0.94; P = 0.004) was found between IC50 values obtained using the two MDR-positive cell lines. Conversely, IC50 values obtained in MDA-MB 231 cells did not show a significant correlation with MDR-positive cell lines, thereby suggesting some difference in the antiproliferative mechanism(s) of colchicine analogues. Cell cycle analysis of the most active analogues in breast cancer cells showed a relationship between cell cycle blocking activity and growth inhibition. The most active agents on the MDR-positive MCF7 ADRr cell line, after 24 h of culture, in terms of cell cycle blocking activity were the three fluorinated analogues. Interestingly, after 72 h, when the cell cycle block subsided, a consistent amount of DNA fragmentation was evident. The extent of cell cycle block, measured as the G2/G1 ratio, was significantly correlated with the apoptosis rate expressed as a percentage of DNA fragmentation on both cell lines, thereby suggesting that a large number of blocked cells underwent the apoptotic pathway.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C9754
Colchicine, ≥95% (HPLC), powder
C22H25NO6
SML0954 Thiocolchicine, ≥97% (HPLC)
C22H25NO5S