EMAIL THIS PAGE TO A FRIEND
The New England journal of medicine
Treatment of postmenopausal osteoporosis.
PMID 9494151
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
450022
Sodium fluoride, anhydrous, powder, 99.99% trace metals basis
FNa