EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
MKK6 activates myocardial cell NF-kappaB and inhibits apoptosis in a p38 mitogen-activated protein kinase-dependent manner.
PMID 9525929
Abstract
In cardiac myocytes the stimulation of p38 mitogen-activated protein kinase activates a hypertrophic growth program and the induction of the cardiac-specific genes associated with this program. This study focused on determining whether these novel growth-promoting effects are accompanied by the p38-mediated inhibition of apoptosis, and if so, what signaling pathways might be responsible. Primary neonatal rat ventricular myocytes were driven into apoptosis by treatments known to induce apoptosis in other cell types, e.g. incubation with anisomycin or overexpression constitutively active MEKK-1 (MEKK-1COOH), a protein that strongly activates extracellular signal-regulated kinase and N-terminal c-Jun kinase, but not p38. Overexpression of constitutively active MKK6, MKK6 (Glu), which selectively activates p38 in cardiac myocytes, protected cells from either anisomycin- or MEKK-1COOH-induced apoptosis. This protection was blocked by SB 203580, a selective p38 inhibitor. MKK6 (Glu) also activated transcription mediated by NF-kappaB, a factor which protects other cell types from apoptosis. The activation of NF-kappaB and the protection from apoptosis mediated by MKK6 (Glu) were both blocked by SB 203580. Interestingly, overexpression of a mutant form of I-kappaBalpha, which inhibits nuclear translocation of NF-kappaB, completely blocked MKK6 (Glu)-activated NF-kappaB but had little effect on MKK6s anti-apoptotic effects. These findings suggest that, in part, the overexpression of MKK6 (Glu) may foster growth and survival of cardiac myocytes by protecting them from apoptosis in a p38-dependent manner. Additionally, while NF-kappaB is activated in myocardial cells by p38, this does not appear to be the major mechanism by which MKK6 (Glu) exerts its anti-apoptotic effects in this cell type, suggesting a novel pathway for p38-mediated protection from apoptosis.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A9789
Anisomycin from Streptomyces griseolus, ≥98% (HPLC), solid
C14H19NO4