EMAIL THIS PAGE TO A FRIEND
Nature genetics
Neurofibromatosis 2 tumour suppressor schwannomin interacts with betaII-spectrin.
PMID 9537418
Abstract
NF2 is the most commonly mutated gene in benign tumours of the human nervous system. The NF2 protein, called schwannomin or merlin, is absent in virtually all schwannomas, and many meningiomas and ependymomas. Using the yeast two-hybrid system, we identified betaII-spectrin (also known as fodrin) as a schwannomin-binding protein. Interaction occurred between the carboxy-terminal domain of schwannomin isoform 2 and the ankyrin-binding region of betaII-spectrin. Isoform 1 of schwannomin, in contrast, interacted weakly with betaII-spectrin, presumably because of its strong self-interaction. Thus, alternative splicing of NF2 may regulate betaII-spectrin binding. Schwannomin co-immunoprecipitated with betaII-spectrin at physiological concentrations. The two proteins interacted in vitro and co-localized in several target tissues and in STS26T cells. Three naturally occurring NF2 missense mutations showed reduced, but not absent, betaII-spectrin binding, suggesting an explanation for the milder phenotypes seen in patients with missense mutations. STS26T cells treated with NF2 antisense oligonucleotides showed alterations of the actin cytoskeleton. Schwannomin itself lacks the actin binding sites found in ezrin, radixin and moesin, suggesting that signalling to the actin cytoskeleton occurs via actin-binding sites on betaII-spectrin. Thus, schwannomin is a tumour suppressor directly involved in actin-cytoskeleton organization, which suggests that alterations in the cytoskeleton are an early event in the pathogenesis of some tumour types.